Ultra One® Latex Gloves, Microflex®, Ansell
Proveedor: ANSELL HEALTHCARE PRODUCTS LLC MX
Certificados
Acerca de este artículo
These gloves have extended, beaded cuffs for increased protection over the wrist and forearm.
- A standard for high-risk applications
- Fingertip texture allows for better grip in wet and dry conditions
Embalaje: Packaged 50 gloves per dispenser box.
Precaución: This product contains natural rubber latex which may cause allergic reactions. Safe use of this glove by or on latex sensitized individuals has not been established.
Especificaciones
- Catalog No:
- 32916-516
- 32916-518
- 32916-520
- Ambidiestro/Específico para cada mano:
- Ambidiestro
- Ambidiestro
- Ambidiestro
- Desechable / Reutilizable:
- Desechable
- Desechable
- Desechable
- Latex/No contiene Latex:
- Latex
- Latex
- Latex
- Con polvo/sin polvo:
- Sin polvo
- Sin polvo
- Sin polvo
- Espesor del puño:
- 7.1 mil
- 7.1 mil
- 7.1 mil
- Espesor del dedo:
- 13.8 mil
- 13.8 mil
- 13.8 mil
- Espesor de la palma:
- 9.8 mil
- 9.8 mil
- 9.8 mil