CE4 System Latex Ambidextrous Gloves, Microflex®, Ansell
Supplier: ANSELL HEALTHCARE PRODUCTS LLC MX
Certificates
About this item
These ambidextrous gloves are packaged with a double vacuum seal for use in ISO Class 4 (FED-STD-209E Class 10/M2.5) controlled environments.
- Silicone-free, powder-free
- Microtextured surface enhances wet or dry grip
- Provides superb aesthetics, dexterity, tactile sense, fit, feel, and comfort
These gloves are washed in microprocessor-controlled, two-in-one, washer/dryer laundering systems to provide very low surface particulates, ionic extractables, and non-volatile residues.
Certifications: Lot-specific Certificate of Analysis available upon request.
Packaging: Packed 100 gloves per vacuum-sealed double poly bag, 10 poly bags per case.
Caution: These gloves contain natural rubber latex which may cause allergic reactions. Safe use of this glove by or on latex-sensitized individuals has not been established.
Specifications
- Catalog No:
- 32916-720
- 32916-718
- Ambidiestro/Específico para cada mano:
- Ambidiestro
- Ambidiestro
- Latex/No contiene Latex:
- Latex
- Latex
- Con polvo/sin polvo:
- Sin polvo
- Sin polvo
- Espesor del dedo:
- 7.9 mil
- 7.9 mil
- Espesor de la palma:
- 6.3 mil
- 6.3 mil