Orders LinkContactUsLinkComponent
CE5 System Latex Gloves, Microflex®, Ansell
CE5 System Latex Gloves, Microflex®, Ansell
CE5 System Latex Gloves, Microflex®, Ansell
Certificates
CE5 System Latex Gloves, Microflex®, Ansell
1 Options Available Below
Product Details & Documents
These ambidextrous gloves are for use in ISO Class 5 (FED-STD-209E Class 100/M3.5) controlled environments.
  • Low surface particulates, ionic extractables, and non-volatile residues
  • Silicone-free
  • Micro-textured surface enhances wet or dry grip
  • Provides superb aesthetics, dexterity, tactile sense, fit, feel, and comfort
  • Powder-free
Textured, ambidextrous gloves are compatible for use with both wet and dry environments.
 : Lot-specific Certificate of Analysis available upon request.
 : Packed 100 gloves per vacuum-sealed double poly bag, 10 poly bags per case.
 : These gloves contain natural rubber latex which may cause allergic reactions. Safe use of this glove by or on latex-sensitized individuals has not been established.
Specifications for Product Family
Specifications
  • Catalog No:
  • 32916-712
  • 32916-714
  • Ambidextrous / hand-specific:
  • Ambidextrous
  • Ambidextrous
  • Latex / latex-free:
  • Latex
  • Latex
  • Powdered / powder-free:
  • Powder-free
  • Powder-free
  • Finger thickness:
  • 7.9 mil
  • 7.9 mil
  • Palm thickness:
  • 6.3 mil
  • 6.3 mil
Specifications
Product Family Options
Product Information
  • Glove size
    Glove Color
    Length
    Availability
    Price
  • M
    Natural
    30.5 cm (12")
    Discontinued
    Supplier Number: CE5-512-M
    Specifications:
    More
    Specifications
    Cat. No.
    32916-712
    Ambidextrous / hand-specific
    Ambidextrous
    Latex / latex-free
    Latex
    Powdered / powder-free
    Powder-free
    Finger thickness
    7.9 mil
    Palm thickness
    6.3 mil
  • L
    Natural
    30.5 cm (12")
    Log in for availability
    Log in for availability
    Case (1,000UN)
    Case (1,000UN) - $9,090.00
    $9,090.00
    Case (1,000UN)
       $9,090.00
    Supplier Number: CE5-512-L
    Specifications:
    More
    Specifications
    Cat. No.
    32916-714
    Ambidextrous / hand-specific
    Ambidextrous
    Latex / latex-free
    Latex
    Powdered / powder-free
    Powder-free
    Finger thickness
    7.9 mil
    Palm thickness
    6.3 mil