Orders LinkContactUsLinkComponent
VWR® Flask and Bottle Brushes, Spring End
VWR® Flask and Bottle Brushes, Spring End
VWR® Flask and Bottle Brushes, Spring End
VWR® Flask and Bottle Brushes, Spring End
White, natural bristles are mounted on a flexible plastic handle.
  • Flexible plastic handle
  • Narrow shaft easily fits deep inside larger flasks and bottles
  • Ideal for cleaning volumetric flasks
Design ideal for cleaning volumetric flasks. Angled handle flexes conveniently to reach past narrow necks.
Product Family Options
  • Description
    Bristle Material
    Brush Head Length
    Handle Material
    Total length
  • For 250 mL flasks
    Hog hair
    8.9 cm (3 1/2")
    Plastic
    30.5 cm (12")
    Paquete de 5
    Paquete de 5 - $2,080.32
    $2,080.32
    Paquete de 5
       $2,080.32
  • For 500 mL flasks
    Hog hair
    10.2 cm (4")
    Plastic
    35.6 cm (14")
    Paquete de 5
    Paquete de 5 - $2,278.44
    $2,278.44
    Paquete de 5
       $2,278.44
  • For 1000 mL flasks
    Hog hair
    12.7 cm (5")
    Plastic
    43 cm (17")
    Paquete de 5
    Paquete de 5 - $2,501.34
    $2,501.34
    Paquete de 5
       $2,501.34