ContactUsLinkComponent
KIMAX® Color-Coded Griffin Beakers, Double Scale, Borosilicate Glass
  89000-912
 : 14000Y-100
undefined
KIMAX® Color-Coded Griffin Beakers, Double Scale, Borosilicate Glass
  89000-912
 : 14000Y-100
Select an Option
50 mm
Select an Option
10 ml
Select an Option
20 - 80 ml
Select an Option
100 ml (3.4 oz.)
Select an Option
71 mm
Select an Option
89000-912
Select an Option
14000Y-100
These low-form borosilicate glass beakers feature brightly screened marking areas and graduations in blue, green, red, or yellow.
  • Durable matte finish of marking area allows use of a pencil
  • Double-graduated scale is easy to read
Designed from ASTM Specifications E960, Type I requirements.
Frequently Bought Together