Orders LinkContactUsLinkComponent
PYREX® Glass Beads, Corning
3 mm
  89091-366
 :   Corning
 :  7268-3
PYREX® Glass Beads, Corning
  89091-366
 :  Corning
PYREX® Glass Beads, Corning
PYREX® Glass Beads, Corning
3 mm
$45,830.00
Case (4UN)
Solid glass.
Glass beads used as packing for distillation columns, mixing beads and boiling stones.
Supplied in a 0.45 kg (1 lb.) box with a packing volume of approximately 360 cm³ (22 cu. in.).
  • Ø:
    3 mm