VWR® Dropping Bottles, PTFE
25 ml (0.85 oz.), 33×80 mm
Catalog # 89026-086
Supplier: VWR International
Jump to:
Product Details & Documents
PTFE dropping bottles are suitable for dispensing and dropping small volumes of reagents and aggressive chemicals.
- Chemically resistant and inert
- Tight, leak-free seal
- Bottle sides are flexible
Bottle sides are flexible to enable user to squeeze out contents. Screw-on cap ensures a tight, leak-free seal.
Especificaciones
- Capacity:25 ml (0.85 oz.)
- Diameter ×H:33×80 mm