Orders LinkContactUsLinkComponent
VWR® Digital Anemometer with Thermometer
VWR® Digital Anemometer with Thermometer
Anemometer with thermometer, 1.2 m (4 ft.) cable
Catalog # 33500-090
Supplier:   VWR International
VWR® Digital Anemometer with Thermometer
Catalog # 33500-090
Supplier:  VWR International
VWR® Digital Anemometer with Thermometer
VWR® Digital Anemometer with Thermometer
Anemometer with thermometer, 1.2 m (4 ft.) cable
Price
$11,358.33
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
Device measures air velocity and temperature in hoods and outdoor environments with ease and accuracy.
  • Traceable®
Low-friction ball bearing design allows the vane to rotate freely at low and high velocities to register even slight gas movement in ducts
Rugged construction, including a chemical- and shock-resistant ABS plastic case, is suitable for heavy use in lab, plant, and field. An extra-large, 1.9 cm (3/4") high LCD, legible from 4.6m (15'), and built-in, flip-open bench stand facilitate easy monitoring. Ease of operation ensures consistent results.
Readings are updated instantaneously. Memory function records highest and lowest readings. Hold switch freezes the display at current reading, which is ideal for capturing a reading in a difficult location.
Certifications: A certificate is provided to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: Supplied with AAA alkaline battery and carrying case.
Especificaciones
  • Description:
    Anemometer with thermometer, 1.2 m (4 ft.) cable
  • Cat. No.:
    33500-090
  • Speed resolution:
    0.1 m/sec., 0.1 km/hr., 10 ft./min., or 0.1 knots
  • Speed range:
    0.4–30.0 m/sec., 1.4–108 km/hr., 80–5910 ft./min., or 0.8–53.3 nautical mph
  • Speed accuracy:
    ±(2% + 1 digit)
  • Dimensions:
    6.4H x 16.5W x 3.2D cm (21/2 x 61/2 x 11/4")
  • Temperature range:
    0 to 60°C (32 to 140°F)
  • Temperature resolution:
    0.1°
  • Temperature accuracy:
    ±1°C
  • Weight:
    0.45 kg (1 lb.)