Orders LinkContactUsLinkComponent
Modular Storage Cabinets, Akro-Mils™
Modular Storage Cabinets, Akro-Mils™
9-Drawer with Steel Door and Lock, 7.8H×13.2W×26.8D cm (31/16×53/16×109/16"); Frame dimensions: 43W×28H×28D cm (17×11×11")
Catalog # 18700-146
Supplier:   AKRO-MILS
Número de proveedor #:  19-109
Modular Storage Cabinets, Akro-Mils™
Catalog # 18700-146
Supplier:  AKRO-MILS
Modular Storage Cabinets, Akro-Mils™
Modular Storage Cabinets, Akro-Mils™
9-Drawer with Steel Door and Lock, 7.8H×13.2W×26.8D cm (31/16×53/16×109/16"); Frame dimensions: 43W×28H×28D cm (17×11×11")
Price
$5,126.31
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
Perfect for storing clamps, stirring bars, filters, connectors, and more
For valuable or restricted items, model 18700-146 features a steel door and lock that limit access and provide security.
Heavy-duty, 16-gauge steel modular cabinets are designed with steel shelves and center posts. Units mount on any wall using back keyholes, or stack using specially designed nesting feet. Charcoal gray, baked-enamel finish. High-impact polystyrene drawers are corrosion-resistant. Transparent windows and label holders facilitate storage and identification.
 : Shipped complete with frame, drawers, and drawer dividers. Locking model 18700-146 is supplied with two keys.
Especificaciones
  • Description:
    9-Drawer with Steel Door and Lock
  • Dimensions:
    7.8H×13.2W×26.8D cm (31/16×53/16×109/16"); Frame dimensions: 43W×28H×28D cm (17×11×11")