SP Bel-Art Indicator Bottles, LDPE, Bel-Art Products, a part of SP
Catalog # 16354-068
Supplier: Bel-Art Products, a Part of SP
Specifications
- Capacidad:60 ml (2 oz.)
- Altura:9.4 cm (3.7")
- Referencia:16354-068
- Nº de proveedor:116620000
Specifications
About this item
Ideal for use with indicator solutions or dyes.
- LDPE constructoin
- Removable dropper
- Can be used as spray bottle or dropper bottle
Use as a spray bottle if used with the supplied tube or a dropper bottle with the tube removed. Dropper removable for filling. With polypropylene screw cap, polyethylene dropper attachment and tube.