ContactUsLinkComponent
KIMAX® Griffin Beakers, Low Form, Double Scale, Borosilicate Glass, Kimble Chase
KIMAX® Griffin Beakers, Low Form, Double Scale, Borosilicate Glass, Kimble Chase
Catalog # 89001-060
Supplier #: 14000-150
undefined
KIMAX® Griffin Beakers, Low Form, Double Scale, Borosilicate Glass, Kimble Chase
Catalog # 89001-060
Supplier Number:  14000 30
Supplier #: 14000-150
Log in for availability
Select an Option
20 ml
Select an Option
20 - 140 ml
Select an Option
150 ml (5 oz.)
Select an Option
89001-060
Select an Option
14000-150
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
About this item
These Griffin beakers feature thick, slightly flared, beaded tops with a spout designed for excellent pouring characteristics.
  • Durable matte finish marking area
  • White graduated scale for measuring and/or mixing where a high level of accuracy is not required
They have uniform walls throughout and are constructed of KIMAX KG-33 glass.
Certifications: Designed from ASTM Specification E960, Type I requirements.
Frequently Bought Together