ContactUsLinkComponent
VITLAB® Griffin Beakers, PMP, BrandTech
VITLAB® Griffin Beakers, PMP, BrandTech
Catalog # 83008-800
Supplier #: 61003
undefined
VITLAB® Griffin Beakers, PMP, BrandTech
Catalog # 83008-800
Supplier Number:  60503
Supplier #: 61003
Log in for availability
Select an Option
77 mm
Select an Option
25 ml
Select an Option
250 ml (8.5 oz.)
Select an Option
95 mm (3.74")
Select an Option
83008-800
Select an Option
V61003
About this item
High-quality beakers made of plastic polymethylpentene with moderately good chemical resistance.
  • Glass-clear polymethyl pentene (PMP)
  • Red screened graduations for high contrast with many liquids
Beakers feature molded graduations with red-screened overprint for legibility.
Certifications: All beakers meet the requirements of ISO 7056.
Frequently Bought Together