Nalgene® Polypropylene Copolymer Dilution Bottles, Thermo Scientific
Catálogo # 16161-767
Proveedor: Thermo Fisher Scientific
Especificaciones
- Tamaño de la tapa:38-415
- Capacidad:205 ml (6.9 oz.)
- Referencia:16161-767
- Estilo:Wide Mouth
- Nº de proveedor:2505-0380
Especificaciones
Acerca de este artículo
Lightweight, break-resistant, autoclavable bottles for preparing serial dilutions in quantitative microbiological analyses.
- Useful temperature range of –40 to 121 °C (–40 to 250 °F)
- Accurate to ±4 ml
Narrow mouth (28 mm) bottle is intended for liquid samples; wide mouth (38 mm) is designed for solid or liquid samples. Molded-in graduations at both 90 and 99 ml for easy preparation of ¹/₁₀ or ¹/₁₀₀ dilutions in the same bottle.
Contact-clear bottles have excellent resistance to strong acids, alcohols, and bases. Withstand repeated autoclaving and washing with Tween®. Can also be used as culture vessels, sampling, or storage bottles.
For replacement caps, see 16219-030 series.
Información de pedido: Supplied with linerless polypropylene screw closure, guaranteed leakproof. Bottles available in narrow mouth and wide mouth styles.
Precaución: Before autoclaving, set cap on top of bottle without engaging threads.