ContactUsLinkComponent
VWR®, Graduated Beakers with Handle, Low Form, Polypropylene
VWR®, Graduated Beakers with Handle, Low Form, Polypropylene
Catalog # 13916-730
Supplier:  VWR International
undefined
VWR®, Graduated Beakers with Handle, Low Form, Polypropylene
Catalog # 13916-730
Supplier:  VWR International
Log in for availability
Select an Option
50 - 1000 ml
Select an Option
1 L (34 oz.)
Select an Option
13916-730
About this item
Graduated beakers are robust, with a non-drip spout and ridged thumb-grip on handle.
  • Molded of autoclavable, translucent polypropylene
  • Low form for better stability
  • All sizes graduated in milliliters and liters
Frequently Bought Together