ContactUsLinkComponent
SP Bel-Art Tall Form Graduated Pitchers, Polypropylene, Bel-Art Products, a part of SP
SP Bel-Art Tall Form Graduated Pitchers, Polypropylene, Bel-Art Products, a part of SP
Catálogo # 53600-265
Número de proveedor #: F28994-0000
undefined
SP Bel-Art Tall Form Graduated Pitchers, Polypropylene, Bel-Art Products, a part of SP
Catálogo # 53600-265
Número de proveedor:  F28995-0000
Número de proveedor #: F28994-0000
Iniciar sesión Para disponibilidad
Select an Option
152 mm (6") Bottom; 215 mm (8<sup>1</sup>/<sub>2</sub>") Top
Select an Option
100 ml
Select an Option
5000 ml (169 oz.)
Select an Option
270 mm (10⅜")
Select an Option
53600-265
Select an Option
289940000
Acerca de este artículo
Available in a large selection of sizes, these pitchers are ideal for general laboratory use and offer well designed spouts and large open handles for easy stacking.
  • Use with wet or dry ingredients
  • Chemical resistant and shatterproof
  • Translucent polypropylene construction
  • Heat resistant to 121 ºC (250 ºF); steam autoclavable
Metric gradations molded on outside wall.
Frequently Bought Together