Nalgene® Griffin Low Form Graduated Beakers, Polypropylene, Thermo Scientific
13915-067
: 1201-0030
Low Manufacturing Impact
Product Transparency and Disclosure
- Catalog No:
- 13915-067
- 13915-089
- 13915-078
- 13915-103
- 13915-125
- 13915-114
- 13915-169
- 13915-147
- 13915-180
- Environmentally Preferable:
- Low Manufacturing Impact, Product Transparency and Disclosure
- Low Manufacturing Impact, Product Transparency and Disclosure
- Low Manufacturing Impact, Product Transparency and Disclosure
- Low Manufacturing Impact, Product Transparency and Disclosure
- Low Manufacturing Impact, Product Transparency and Disclosure
- Low Manufacturing Impact, Product Transparency and Disclosure
- Low Manufacturing Impact, Product Transparency and Disclosure
- Low Manufacturing Impact, Product Transparency and Disclosure
- Low Manufacturing Impact, Product Transparency and Disclosure
- Certificación ambiental:
- ISO14001 (Certificate Registration No. EMS 752603), ACT Label
- ISO14001 (Certificate Registration No. EMS 752603), ACT Label
- ISO14001 (Certificate Registration No. EMS 752603), ACT Label
- ISO14001 (Certificate Registration No. EMS 752603), ACT Label
- ISO14001 (Certificate Registration No. EMS 752603), ACT Label
- ISO14001 (Certificate Registration No. EMS 752603), ACT Label
- ISO14001 (Certificate Registration No. EMS 752603), ACT Label
- ISO14001 (Certificate Registration No. EMS 752603), ACT Label
- ISO14001 (Certificate Registration No. EMS 752603), ACT Label
Heavy-duty, translucent polypropylene beakers with ribs for stacking.
- Easy-to-read graduations
- Autoclavable
- Chemically resistant
Beakers feature easy-to-read graduations with size and resin codes. Maximum use temperature and no-flame symbol printed in black. Autoclavable and chemically resistant. Not for use on a hot plate.
Frequently Bought Together