ContactUsLinkComponent
PYREX® Heavy-Duty Griffin Beakers, Graduated
PYREX® Heavy-Duty Griffin Beakers, Graduated
Catalog # 13912-524
Supplier:  Corning
   Sustainable Options Available
Supplier #: 1003-250
Product available on GSA Advantage®
undefined
PYREX® Heavy-Duty Griffin Beakers, Graduated
Catalog # 13912-524
Supplier:  Corning
Supplier Number:  1003-150
   Sustainable Options Available
Supplier #: 1003-250
Product available on GSA Advantage®
Log in for availability
Select an Option
25 - 200 ml
Select an Option
250 ml (8.5 oz.)
Select an Option
13912-524
Select an Option
1003-250
Specifications
  • Catalog No:
  • 13912-524
  • 13912-546
  • 89090-528
  • 89090-526
  • 13912-604
  • 13912-502
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Certificación ambiental:
  • I-RECs, ISO 14001 - Monterrey
Specifications
About this item
PYREX® Heavy-Duty Griffin beakers are made of PYREX® borosilicate glass.
  • Very good chemical resistance
  • Thermal shock resistance
  • With extra large marking spot
Heavy-duty beakers are specifically designed to offer the best mechanical strength under harsh conditions, such as mechanized washing operations. PYREX® Heavy-Duty Griffin beakers in sizes 250 ml and larger have a double graduated metric scale to indicate approximate content.
Certifications: Meet ASTM E960 specifications.
Frequently Bought Together