Orders LinkContactUsLinkComponent
Cryogenic Storage Boxes, Polycarbonate, Corning®
Cryogenic Storage Boxes, Polycarbonate, Corning®
81, 1 - 2 ml, 5/Pack, 10/Case
Catalog # 22250-106
Supplier:   Corning
Número de proveedor #:  431119
Cryogenic Storage Boxes, Polycarbonate, Corning®
Catalog # 22250-106
Supplier:  Corning
Cryogenic Storage Boxes, Polycarbonate, Corning®
Cryogenic Storage Boxes, Polycarbonate, Corning®
81, 1 - 2 ml, 5/Pack, 10/Case
Price
$2,150.00
Log in Para disponibilidad
Paquete de 5 Case de 2 (5UN)
Product Details & Documents
These cryogenic storage boxes are suitable for storage down to –196°C (–320.8°F). These boxes feature a large marking spot and a clear polycarbonate lid that can be labeled to show vial contents. They are stackable and are available in two formats: 81-place boxes which will hold internally and externally threaded vials; and 100 count boxes, which will hold internally threaded vials.
  • Reusable
  • Ridges on the sides of the box make them easier to hold while wearing gloves
  • Transparent lid with numeric grid
Designed for use with most cryogenic vials.
These boxes feature a large marking spot and a clear polycarbonate lid that can be labeled to show vial contents. They are stackable and are available in two formats: 81-place boxes which will hold internally and externally threaded vials; and 100 count boxes, which will hold internally threaded vials.
These cryogenic storage boxes are suitable for storage down to –196°C (–320.8°F).
Documents
Certificates
Buscar por número de lote
Especificaciones
  • No. of Holes:
    81
  • Size:
    5/Pack, 10/Case
  • For Vials:
    1 - 2 ml