Orders LinkContactUsLinkComponent
VWR®, Beaker Cups, Polystyrene
VWR®, Beaker Cups, Polystyrene
Supplier:  VWR International
Certificates
VWR®, Beaker Cups, Polystyrene
VWR®, Beaker Cups, Polystyrene
4 Options Available Below
Product Details & Documents
Beaker cups are designed for use with various automated procedures and as an economical, general-purpose sample cup.
  • Disposable
  • Polystyrene fabrication
  • Economical sample cup
The 5 mL and 20 mL sizes are recommended for flame photometry analysis.
Product Family Options
Información del producto
  • Capacity
    Availability
    Price
  • 5 ml (0.17 oz.)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (1,000UN)
    Case (1,000UN) - $679.75
    $679.75
    Case (1,000UN)
       $679.75
  • 10 ml (0.34 oz.)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (1,000UN)
    Case (1,000UN) - $736.39
    $736.39
    Case (1,000UN)
       $736.39
  • 20 ml (0.68 oz.)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (500UN)
    Case (500UN) - $639.30
    $639.30
    Case (500UN)
       $639.30
  • 150 ml (5 oz.)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (100UN)
    Case (100UN) - $442.61
    $442.61
    Log in Para disponibilidad
    Each (100UN)
    Each (100UN) - $442.61
    $442.61
    Case (100UN)
       $442.61
    Each (100UN)
       $442.61