ContactUsLinkComponent
Nalgene® Dispensing and Storage Jugs, Polypropylene, Thermo Scientific
  16093-011
undefined
Nalgene® Dispensing and Storage Jugs, Polypropylene, Thermo Scientific
  16093-011
 :  2221-0010
  • Tamaño de la tapa:
    38-430
  • Capacidad:
    4 L (1 gal.)
  • Referencia:
    16093-011
  • Nº de proveedor:
    2221-0010
Autoclavable, leakproof jugs offer excellent chemical resistance.
  • Leakproof
  • Autoclavable
Note: Before autoclaving, set cap on top of jug without engaging the threads.
A large carrying handle allows for the use of rubber gloves for safe handling of corrosive liquids, as well as an offset pour spout for easy, accurate pouring.
 : Jugs are supplied with polypropylene screw closures. For replacement caps, see 16219-122 series