Nunc® Vial Storage Boxes, Thermo Scientific
55710-257
- Núm. de orificios:81
- Color:White
- Dimensiones:132L×132W×51H mm (53/16×53/16×2")
- Referencia:55710-257
- For Vials:1.0 - 1.8 ml
- Nº de proveedor:378220
Polyethylene-coated chipboard boxes are vented for good drainage and fit standard rack systems. Supplied with dividers.
Supplied with dividers