Orders LinkContactUsLinkComponent
Nalgene® Rectangular Carboys with Handle, Polypropylene, Thermo Scientific
Nalgene® Rectangular Carboys with Handle, Polypropylene, Thermo Scientific
Nalgene® Rectangular Carboys with Handle, Polypropylene, Thermo Scientific
Certificates
Nalgene® Rectangular Carboys with Handle, Polypropylene, Thermo Scientific
Nalgene® Rectangular Carboys with Handle, Polypropylene, Thermo Scientific
2 Options Available Below
Product Details & Documents
Ideal for storing solutions, media, and sterile water.
  • Leakproof
For replacement caps, see cap 16219-106 in 16219-030 series.
Note: Before autoclaving, set cap on top of carboy without engaging threads.
Useful for handling large volumes of powders and other solid samples. Wide mouth allows easy, efficient cleaning. Space-saving carboys molded in tough, autoclavable polypropylene with 8.9 cm (3¹/₂") neck opening. Sturdy handle attached to molded-in shoulder loops. Calibrated in liters and gallons. Cap size: 100-415.
Documents
Product Family Options
Product Information
  • Capacity
    Dimensions
    Availability
    Price
  • 9 L (2.4 gal.)
    21.6 x 14.6 x 35.6 cm (81/2 x 53/4 x 14")
    Log in for availability
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $1,811.61
    $1,811.61
    Log in for availability
    Case de 6 (1UN)
    Case de 6 (1UN) - $10,869.67
    $10,869.67
    Each (1UN)
       $1,811.61
    Case de 6 (1UN)
       $10,869.67
    Supplier Number: 2212-0020
  • 20 L (5.3 gal.)
    31.8 x 21.9 x 38.1 cm (121/2 x 85/8 x 15")
    Log in for availability
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $6,150.00
    $6,150.00
    Log in for availability
    Case de 4 (1UN)
    Case de 4 (1UN) - $24,784.76
    $24,784.76
    Each (1UN)
       $6,150.00
    Case de 4 (1UN)
       $24,784.76
    Supplier Number: 2212-0050