Sash Brush, Gordon Brush
Hog hair, 1 1/8"), Plastic, 9 1/2", 4.8 cm (1⅞"), #4
Catalog # 22940-748
Supplier: GORDON BRUSH MFG CO
Número de proveedor #: 900046
Jump to:
Product Details & Documents
This Size 4 hog bristle and plastic handle sash brush has an oval shape, and is excellent for dusting and removing debris.
- Metal ferrule
- Large tapered plastic handle
This useful brush has a trim length of 1-7/8", an overall length of 9-1/2" x 3/4" and a brush face width of 1-1/8".
Especificaciones
- Bristle Material:Hog hair
- Handle Material:Plastic
- Size:#4
- Total length:9 1/2"
- Brush Head Width:1 1/8")
- Trim Length:4.8 cm (1⅞")