Chemware® PTFE Boiling Stones, Saint-Gobain Performance Plastics
26397-103
- Descripción:PTFE boiling stones
- Referencia:26397-103
- Empaquetado:450 g
- Nº de proveedor:D1069103
Ultra-pure, virgin PTFE chips.
- promote gentle, efficient boiling by creating many projections on which small bubbles can form
- Unaffected by boiling acids, hydrocarbons, ketones, esters, alcohols, or alkalies in any concentrations
- Suitable for column packing
Ideal for continuous extraction methods, such as EPA SW846.
: Supplied in a 450 g bottle.