Paint/Dust Brushes, Mill-Rose
Natural, 2.5 cm (1"), Wood, 20.7 cm (77/8")
Catalog # 17235-020
Supplier: MILL-ROSE COMPANY
Supplier #: 75117B
Jump to:
Product Details & Documents
Washable paint/dust brushes are just right for dusting balances and bench surfaces.
Soft black natural bristles and wooden handle are held together with a nailed metal ferrule.
Specifications
- Bristle Color:Natural
- Handle Material:Wood
- Total length:20.7 cm (77/8")
- Brush Head Width:2.5 cm (1")