Orders LinkContactUsLinkComponent
Hard Rubber Spatulas
Hard Rubber Spatulas
Hard Rubber Spatulas
Supplier:  JOHN M. MARIS CO
Certificates
Hard Rubber Spatulas
Hard Rubber Spatulas
3 Options Available Below
Product Details & Documents
Ideal for use with materials that affect metals adversely, for mixing, weighing, cleaning, blending, and many other uses.
  • Non sparking
  • Wide and flexible blade
Hand-finished and hand-polished blades. Spatulas are rust-proof, washable, and sterilizable.
Documents
Product Family Options
Product Information
  • Blade Length
    Availability
    Price
  • 102 mm (4")
    Log in for availability
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $467.98
    $467.98
    Log in for availability
    Paquete de 12 (1UN)
    Paquete de 12 (1UN) - $510.09
    $510.09
    Each (1UN)
       $467.98
    Paquete de 12 (1UN)
       $510.09
    Supplier Number: H/R-4"
  • 152 mm (6")
    Log in for availability
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $514.40
    $514.40
    Log in for availability
    Paquete de 12 (1UN)
    Paquete de 12 (1UN) - $560.70
    $560.70
    Each (1UN)
       $514.40
    Paquete de 12 (1UN)
       $560.70
    Supplier Number: H/R-6"
  • 203 mm (8")
    Log in for availability
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $614.34
    $614.34
    Log in for availability
    Paquete de 12 (1UN)
    Paquete de 12 (1UN) - $669.64
    $669.64
    Each (1UN)
       $614.34
    Paquete de 12 (1UN)
       $669.64
    Supplier Number: H/R-8"