Orders LinkContactUsLinkComponent
VWR® Scoop, PTFE
80L x 30W x 20D mm (31/8x11/8x3/4")
Catalog # 89026-220
Supplier:   VWR International
VWR® Scoop, PTFE
Catalog # 89026-220
Supplier:  VWR International
VWR® Scoop, PTFE
VWR® Scoop, PTFE
80L x 30W x 20D mm (31/8x11/8x3/4")
Price
$1,470.04
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
Non-contaminating PTFE scoop useful for transferring frozen substances and other non-liquids.
  • Chemically inert
  • Non-contaminating
  • PTFE construction
Scoop is suitable for use with frozen substances and other solids.
Especificaciones
  • Dimensions:
    80L x 30W x 20D mm (31/8x11/8x3/4")