Orders LinkContactUsLinkComponent
VWR® Tissue Forceps
VWR® Tissue Forceps
152 mm (6")
Catalog # 82027-446
Supplier:   VWR International
VWR® Tissue Forceps
Catalog # 82027-446
Supplier:  VWR International
VWR® Tissue Forceps
VWR® Tissue Forceps
152 mm (6")
Price
$115.39
Log in Para disponibilidad
Each (1UN) Paquete de 12 (1UN) Case de 60 (1UN)
Product Details & Documents
Tissue forceps are specially designed to grasp tissue with minimal damage to the organic material.
  • Stainless steel construction
  • Straight tips
  • Easy to use
These tools are available in a variety of designs, lengths, and specific uses for laboratory procedures.
Documents
Especificaciones
  • Length:
    152 mm (6")