VWR® Spatula, PTFE-Coated
82027-534
- Longitud:17.8 cm (7")
- Referencia:82027-534
Stainless steel spatula features one rounded end and one flat end, with PTFE coating throughout.
- Stainless steel construction
- PTFE coating
- Nonsticking and non-corrosive
It can be used to transfer corrosive or sticky materials without scratching or damaging container surfaces.