Orders LinkContactUsLinkComponent
VWR® Hemostatic Forceps, Serrated Tip
VWR® Hemostatic Forceps, Serrated Tip
140 mm (5½")
Catalog # 82027-416
Supplier:   VWR International
VWR® Hemostatic Forceps, Serrated Tip
Catalog # 82027-416
Supplier:  VWR International
VWR® Hemostatic Forceps, Serrated Tip
VWR® Hemostatic Forceps, Serrated Tip
140 mm (5½")
Price
$174.12
Log in Para disponibilidad
Each (1UN) Paquete de 10 (1UN) Case de 50 (1UN)
Product Details & Documents
Hemostatic forceps are used in many surgical procedures, usually to control bleeding or the flow of liquids in tubing.
  • Stainless steel fabrication
  • Serrated tips
These forceps have ring handles just like scissors, and ratcheted handles can be locked in multiple positions in order to maintain variable levels of constant pressure.
Especificaciones
  • Length:
    140 mm (5½")