VWR® PTFE-Coated Forceps
114 mm (4½")
Supplier: VWR International
Jump to:
Product Details & Documents
Stainless steel forceps are PTFE-coated.
- Stainless steel construction
- Chemically resistant
- Nonsticking and non-corroding
Forceps are coated with PTFE to imbue forceps with heat and chemical resistance.
Documents
Certificates
Buscar por número de loteEspecificaciones
- Length:114 mm (4½")