VWR® Self-Closing Forceps
127 mm (5")
Catalog # 82027-452
Supplier: VWR International
Jump to:
Product Details & Documents
Stainless steel self-closing forceps feature straight, fine tips and a smooth grip.
- Stainless steel construction
- Fine tips
- Self-closing design
Forceps are passivated and autoclavable.
Especificaciones
- Length:127 mm (5")