Orders LinkContactUsLinkComponent
VWR® Antimagnetic Forceps
114 mm (4½")
  82027-400
VWR® Antimagnetic Forceps
  82027-400
VWR® Antimagnetic Forceps
VWR® Antimagnetic Forceps
114 mm (4½")
$458.85
Each (1UN) Paquete de 12 (1UN) Case de 60 (1UN)
Antimagnetic forceps are constructed from anti-magnetic stainless steel.
  • Good resistance to corrosion
  • Autoclavable
  • For general-purpose use
Forceps feature an overall length of 114 mm (4.5") and are available in a variety of unit sizes, from individual forceps to bulk packages.
  • Length:
    114 mm (4½")