Orders LinkContactUsLinkComponent
Pipe Cleaner Brushes, Mill-Rose
Pipe Cleaner Brushes, Mill-Rose
Supplier:   MILL-ROSE COMPANY
Pipe Cleaner Brushes, Mill-Rose
Supplier:  MILL-ROSE COMPANY
Certificates
Pipe Cleaner Brushes, Mill-Rose
Pipe Cleaner Brushes, Mill-Rose
2 Options Available Below
Product Details & Documents
Continuous lengths of cotton pipe cleaner can be cut as needed for cleaning apertures and narrow tubing.
  • Galvanized wire base provides a strong flexible core
  • For tubes with bottoms, bending the cut end back on itself will help eliminate scratches
For larger bores, a length can be coiled around a finger, tube, or jar for a quick springy brush.
Product Family Options
Información del producto
  • Total length
    Availability
    Price
  • 17227-022
    3 mm (1/8") diameter
    3 mm (1/8") diameter
    15 m (50')
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 1
    Paquete de 1 - $458.45
    $458.45
    Paquete de 1
       $458.45
    Supplier Number: MR51807
  • 17227-044
    5 mm (13/64") diameter
    5 mm (13/64") diameter
    15 m (50')
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 50
    Paquete de 50 - $639.57
    $639.57
    Paquete de 50
       $639.57
    Supplier Number: MR51809