VWR® Gooseneck Forceps
152 mm (6")
Supplier: VWR International
Jump to:
Product Details & Documents
These stainless steel forceps are used for dissecting, handling insects, or positioning specimens for pinning.
- Stainless steel construction
- Serrated tips
The forceps have serrated tips and a handle with a gooseneck shank.
Documents
Especificaciones
- Length:152 mm (6")