Nalgene® Filter Forceps, Thermo Scientific
Straight Tip
Catalog # 28198-976
Supplier: Thermo Fisher Scientific
Supplier #: DS0399-0002
Jump to:
Product Details & Documents
Stainless steel blades have serrated grips for handling membrane filters without risk of damage or contamination
Available in bent tip and straight tip. Bent tip forceps feature X-action design that allows membrane to be held without applying constant pressure. Autoclavable.
Specifications
- Description:Straight Tip