VITLAB® PP Buckets, BrandTech
Catalog # 83009-081
Supplier: BRANDTECH SCIENTIFIC INC.
Specifications
- Capacidad:15 L (3.9 gal.)
- Color:Translucent
- Altura:370 mm (14.5")
- Referencia:83009-081
- Nº de proveedor:V96794
- Diámetro:311 mm (12.25")
Specifications
About this item
Polypropylene (PP) buckets feature a hinged handle, pouring lip, and graduation markings
- PP with hinged handle
- Pouring lip to reduce drips
- No lids available
Excellent chemical resistance.