VWR® Dissecting Forceps, Fine Tip, Curved
Catalog # 82027-406
Supplier: VWR International
Specifications
- Longitud:152 mm (6")
- Referencia:82027-406
Specifications
About this item
VWR® Dissecting Forceps are stainless steel with curved blades and fine points.
- Stainless steel fabrication
- Curved blade with fine points
Dissecting forceps are used for a variety of applications and are available in different lengths.