Orders LinkContactUsLinkComponent
VITLAB® Measuring Scoops, HDPE, BrandTech
VITLAB® Measuring Scoops, HDPE, BrandTech
VITLAB® Measuring Scoops, HDPE, BrandTech
VITLAB® Measuring Scoops, HDPE, BrandTech
One-piece, molded measuring scoops made of high density polyethylene.
  • Eight different sizes
  • High quality
VITLAB® measuring scoops are precision molded from durable high-density polyethylene. Handles are cotoured, with a tapered filling edge. Scoops are food-safe according to EC Directive No. 10/2011.
Product Family Options
  • Capacity
    Length
  • 150 ml (5 oz.)
    25 cm (9⅞")
    Paquete de 12
    Paquete de 12 - $763.00
    $763.00
    Paquete de 12
       $763.00
     : 40493
  • 350 ml (11.8 oz.)
    31 cm (12¼")
    Paquete de 6
    Paquete de 6 - $737.27
    $737.27
    Paquete de 6
       $737.27
     : 40593