SP Bel-Art Non sterile and Sterileware® Sampling Spatulas
Proveedor: Bel-Art Products, a Part of SP
Certificados
Acerca de este artículo
These V-shaped spatulas are ideal for every type of powdered or granulated material.
- Available sterile or non sterile
- Molded from FDA-grade materials
- Safe for food, drug, and cosmetic contact
- V shape is ideal for powdered or granulated material
The sturdy point is perfect for taking sterile samples during process quality inspections and for piercing plastic-lined paper bags or similar packages.
Long 14" spatula is perfect for dislodging caked or crystallized reagents from deep within tall bottles or vessels having a neck diameter as small as 18 mm (³/₄").
These one-piece spatulas are molded from FDA-grade materials and are safe for food, drug, and cosmetic contact.
Sterileware® Spatulas (9 and 14") are made of high-impact polystyrene, individually packaged, sterilized by gamma irradiation, lot stamped, and shipped with a Certificate of Processing. Non sterile 9" spatulas are made of polypropylene.
Packaging: Please note that the sterile packaging bag for 56924-809 was previously 11L×3.5"W, and the new dimensions are 12.5L×5.38"W. Please note that the sterile packaging bag for 89236-110 was previously 16L×5"W and the new dimensions are 12.5L×5.38"W.
Información relacionada
Frequently Bought Together