ContactUsLinkComponent
Nalgene® Buckets, White Polypropylene, Thermo Scientific
Nalgene® Buckets, White Polypropylene, Thermo Scientific
Catálogo # 17306-005
undefined
Nalgene® Buckets, White Polypropylene, Thermo Scientific
Catálogo # 17306-005
Número de proveedor:  7012-0110
Especificaciones
  • Capacidad:
    10.4 L (2.7 gal.)
  • Descripción:
    PP-bucket
  • Referencia:
    17306-005
  • Nº de proveedor:
    7012-0110
Especificaciones
Acerca de este artículo
Crack-resistant polypropylene buckets can be autoclaved or gas sterilized for use in the food industry
Note: Do not sterilize with radiation or dry heat. Not suitable for strong alkalis.
Durable enough for handling photographic chemicals and most corrosive liquids. Nylon-coated wire bail handle. Autoclavable.