Ward's® Safety Bottle Carriers
470352-500
- Capacidad:4.5 L
- Color:Blue
- Descripción:Safety bottle carrier
- Dimensiones:7.2×10.5" (16" height, handle extended)
- Referencia:470352-500
- Peso:2 lbs.
Secure and convenient bottle transport.
- Robust design
- Ergonomic handle prevents hand fatigue and related accidents when carrying heavier loads
- Handle can be pressed down to either side for easier loading and unloading
- Centering ring stabilizes bottle in the center of carrier for safer, steadier transport
- Carriers come in four colors, with designated labeling areas for easy identification
- Made of chemical-resistant thermoplastic elastomer/PP blend
The Safety Bottle Carrier provides a safe and reliable way to transport a variety of bottled reagents in the lab. Available in 1.5 and 4.5 liter capacities, this carrier is the practical answer for any clinical and research laboratory setting.