ContactUsLinkComponent
Science Olympiad 2024 Wind Power Test Apparatus
Science Olympiad 2024 Wind Power Test Apparatus
Catalog # 470217-346
Science Olympiad 2024 Wind Power Test Apparatus
Catalog # 470217-346
Supplier Number:  19-20-558P
Specifications
  • Descripción:
    Hub snap-on for CD, 4 mm
  • Referencia:
    470217-346
  • Nº de proveedor:
    19-20-558P
Specifications
About this item
Prepare for upcoming competitions with this practice kit, the only kits officially licensed and approved by science olympiad.
  • Designed hand-in-hand with the national event supervisors
  • Aligned to NGSS-based science olympiad events
  • 1 pre-assembled test apparatus is included in this kit
  • Event rules can be downloaded FREE from the science olympiad website: soinc.org/rules-2024
This kit features components hand selected by the science olympiad organization.
The 2023 to 2024 science olympiad kits were developed to provide an introduction to each event and are not intended to be an 'out of the box solution' for competition. Students and competitors should practice innovation and creativity to make their design their own competition-ready device/activity.
Ordering information: This wind power test apparatus will come fully assembled, along with (3) extra motors, (2) extra 5 OHM resistors, (2) extra 15 OHM resistors, and (2) extra 25 OHM resistors. This kit is intended to be used for testing Wind Power devices that teams build - it does NOT include the materials or instructions on how to build the actual Wind Power device (*see 2024 science olympiad wind power competition Kit for the device).