Nexus™ Horizontal Clean Bench, 8' Wide
77038-044
- Descripción:8' Nexus™ Horizontal clean bench, 26" deep work area with stainless steel interior, includes base stand
- Modelo:26" deep work area
- Dimensiones:101.0×38.2×50.1"
- Referencia:77038-044
Protects your non-hazardous applications with horizontal unidirectional (laminar) ISO Class 4 air across the work area.
- Efficient unidirectional laminar airflow provided through CAP™ technology
- Intelligent 5" touchscreen interface features intuitive monitoring capabilities
- Reliable with long-life HEPA filters and maximum cleanability that stands up to disinfectants
The Nexus Horizontal clean bench is a laminar flow workstation designed to protect non-hazardous samples and processes from contamination.
The Nexus is the most advanced clean bench on the market. New features include a touchscreen interface, Constant Airflow Profile (CAP™) technology that maintains proper airflow velocity as the HEPA filter loads, the ability to log all operational parameters for compliance with regulatory requirements, clear and detailed alarm and alert notifications, and a full OS with user-customizable features, maintenance logs and diagnostics.
: The 230 V units are CE/UKCA certified.