Vinyl Electrical Tape, Yellow
470043-232
- Descripción:Vinyl electrical tape, yellow
- Referencia:470043-232
- Nº de proveedor:3899YELLOW
Excellent resistance to abrasion, moisture, alkalies, acid, copper corrosion and varying weather conditions.
- ¾" × 60'
- Temflex
Designed to perform in ambient temperatures up to 90 °C (194 °F). The tape is conformable for cold weather application down to 0 °C (32°F).