PREempt™ RTU Disinfectant Wipes, Contec®
Proveedor: CONTEC, INC. MX
Certificados
Acerca de este artículo
One-step disinfectant cleaner wipes utilizing Accelerated Hydrogen Peroxide®. Ideal for critical environments.
- Effective, fast-acting, one-step disinfectant cleaner
- Convenient meltblown polypropylene presaturated wipes
- Low residue and compatible with a wide range of materials
- Odor-free for good operator acceptability
- Emits no VOCs and does not require personal protection equipment (PPE)
- Hydrogen peroxide breaks down into water and oxygen for easy disposal and minimal, to no environmental impact
- Not classifed as dangerous goods
PREempt RTU utilizes Accelerated Hydrogen Peroxide® (AHP), a patented synergistic blend of commonly used ingredients, that when combined with low levels of hydrogen peroxide dramatically increase its efficacy and cleaning performance. AHP combines hydrogen peroxide with surfactants, wetting and chelating agents to create an ideal one step cleaner / disinfectant.
Embalaje: Available in a Soft Pack or in a pull top canister.
Información relacionada
Certificados
Buscar por número de lote10822-456_(L)101123VX02 10822-456_011524VX02 10822-456_021423VX03 10822-456_072020VX06 10822-456_072720VX06
Más
VWR Production insight V1
Documentación del producto
Frequently Bought Together