Buckets, Round, BetaMop™ TX7065 / TX7066, Texwipe
Catalog # 10127-110
Supplier: Texwipe
Specifications
- Capacidad:37 L (10 gal.)
- Descripción:Round Stainless Steel Bucket with Casters
- Referencia:10127-110
- Nº de proveedor:TX7066
Specifications
About this item
Made from 100% 304 stainless steel.
- Completely seamless buckets’ design reduces the risk of contamination build up
- Interior volume marking for more accurate solution dilution
- Stainless steel construction provides durability and resistance to cleaning agents
- Easy bucket cleaning due to unique seamless design
- Buckets are autoclave safe, including ones with casters
- Wheels do not leave black markings on floor
- 8 or 10 gallon capacity
Compatible with all cleanroom string mops and most wringers.