Eisco® Balance Weights Set, Stainless Steel
470344-876
- Referencia:470344-876
- Peso:500 g
- Incluye:200 g, 2×100 g, 50 g, 20 g, 2×10 g, 5 g, 2×2 g, 1 g
- No. of Weights:11
Sets of stainless steel balance weights individually stamped and packed in a premium wooden storage block.
- Individually calibrated balance weights in metric grams
- Made from high-quality stainless steel
- Features easy-to-handle, contoured shape
- Each piece is stamped with weight for easy identification
- Supplied in a premium wooden storage block
Sets include the most commonly used balance weights. Each is made from high-grade stainless steel and stamped with its weight. All sets include a polished wooden storage block with a place for each weight.