Masterflex® Nylon Clamps for Sanitary Fittings, Avantor®
Supplier: VWR International
Certificates
About this item
Collared clamp helps prevent unintentional opening.
- All fittings are compatible with Tri-Clamp® fittings
- Excellent chemical resistance; low leachable characteristics
- Environmentally Preferable:
Specifications
- Catalog No:
- MFLX12805-05
- MFLX00424-TN
- MFLX00424-TO
- Sanitary clamp size (port 1):
- 2"
- 1/2"
- 1-1/2"
- Connection type:
- Sanitary
- Sanitary
- Sanitary
- Sterilisation:
- Autoclave, Gamma
- Autoclave, Gamma
- Autoclave, Gamma
- Garantía:
- 1 year
- 1 year
- 1 year