Orders LinkContactUsLinkComponent
Motic BA410E LED Sextuple Compound Microscope
Motic BA410E LED Sextuple Compound Microscope
BA410E trinocular LED sextuple compound microscope
Catalog # 76595-442
Número de proveedor #:  1100100404311
Motic BA410E LED Sextuple Compound Microscope
Catalog # 76595-442
Motic BA410E LED Sextuple Compound Microscope
Motic BA410E LED Sextuple Compound Microscope
BA410E trinocular LED sextuple compound microscope
Price
$143,247.76
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
The BA410E is Motic's flagship model, and it displays the highest level of optical performance. It also allows a profound diagnosis for those in pathology, haematology and cytology who require the best color fidelity. Increased numerical apertures provide an expanded resolution power for a more detailed specimen evaluation.
  • Head Siedentopf type 30° inclined, 360° rotating [trinocular: (light split = ²⁰/₈₀)]
  • Reversed sextuple nosepiece
  • Coaxial coarse and fine focusing adjustment
  • Built in low position rackless stage with right hand control
  • Achromat swing-out condenser N.A. 0.90 with iris diaphragm
Binocular or trinocular research microscope for diverse questions from life sciences, but also from material sciences. The standard EC-H plan achromat objectives can be replaced by fluorite and apochromatic lenses for maximum optical performance. Demanding users benefit from the system’s flexibility of the BA410E construction kit, ready to configure a personal microscope for individual requirements.
Ordering information: Contact your Avantor representative to learn more about this and other motic solutions. All motic products come with a one-year electrical and five-year mechanical warranty.
Especificaciones
  • Description:
    BA410E trinocular LED sextuple compound microscope